Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611883 MIM: 607960 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCCIP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
BCCIP - BRCA2 and CDKN1A interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016567.3 | Intron | NP_057651.1 | ||||
NM_078468.2 | Intron | NP_510868.1 | ||||
NM_078469.2 | Intron | NP_510869.1 |
DHX32 - DEAH-box helicase 32 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018180.2 | Intron | NP_060650.2 | ||||
XM_017016404.1 | Intron | XP_016871893.1 | ||||
XM_017016405.1 | Intron | XP_016871894.1 |
Set Membership: |
HapMap Validated |