Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCTACTCCTCCGTCCCCACTCAC[A/G]TTAGTTAACAGTCCCTCCTCTCTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608541 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP32 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGAP32 - Rho GTPase activating protein 32 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142685.1 | 8473 | UTR 3 | NP_001136157.1 | |||
NM_014715.3 | 8473 | UTR 3 | NP_055530.2 | |||
XM_005271736.4 | 8473 | UTR 3 | XP_005271793.2 | |||
XM_011543072.2 | 8473 | UTR 3 | XP_011541374.1 | |||
XM_011543073.2 | 8473 | UTR 3 | XP_011541375.2 | |||
XM_011543075.2 | 8473 | UTR 3 | XP_011541377.1 | |||
XM_017018595.1 | 8473 | UTR 3 | XP_016874084.1 | |||
XM_017018596.1 | 8473 | UTR 3 | XP_016874085.1 | |||
XM_017018597.1 | 8473 | Intron | XP_016874086.1 |