Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGATGATTGGAAAGGAAGGCCAA[A/C]AGGCAGAGCAGCTAAGGGTGTGGTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606792 MIM: 615319 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HRH4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HRH4 - histamine receptor H4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143828.1 | Intron | NP_001137300.1 | ||||
NM_001160166.1 | Intron | NP_001153638.1 | ||||
NM_021624.3 | Intron | NP_067637.2 | ||||
XM_011526133.1 | Intron | XP_011524435.1 |
IMPACT - impact RWD domain protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |