Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGGAAGATGGGAGGGTCAGGTAT[A/T]TGTCTGCGGGTGGAGAGTCTGCAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600806 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CNN1 - calponin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001299.5 | Intron | NP_001290.2 | ||||
NM_001308341.1 | Intron | NP_001295270.1 | ||||
NM_001308342.1 | Intron | NP_001295271.1 | ||||
XM_005259741.2 | Intron | XP_005259798.1 | ||||
XM_017026289.1 | Intron | XP_016881778.1 |
ELOF1 - elongation factor 1 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |