Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCACGCACTCCGGTCATTCACTCA[C/T]TCATTCATTCATTCATTCACAAAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600806 | ||||||||||||||||||||
Literature Links: |
CNN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CNN1 - calponin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001299.5 | Intron | NP_001290.2 | ||||
NM_001308341.1 | Intron | NP_001295270.1 | ||||
NM_001308342.1 | Intron | NP_001295271.1 | ||||
XM_005259741.2 | Intron | XP_005259798.1 | ||||
XM_017026289.1 | Intron | XP_016881778.1 |
ELOF1 - elongation factor 1 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |