Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGGCAGGGCAGCACCCACAGGTT[A/G]CAGACAGCAATCCTCTTCCACAGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605784 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TTYH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TTYH1 - tweety family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005367.2 | Intron | NP_001005367.1 | ||||
NM_001201461.1 | Intron | NP_001188390.1 | ||||
NM_020659.3 | Intron | NP_065710.1 | ||||
XM_017027025.1 | Intron | XP_016882514.1 | ||||
XM_017027026.1 | Intron | XP_016882515.1 |