Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTTATTTGACTGCTGTTCTCTGCC[A/G]CCTTCCACTCGATGCAAGCTTCTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614790 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
WTIP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WTIP - Wilms tumor 1 interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080436.1 | Intron | NP_001073905.1 | ||||
XM_006723014.3 | Intron | XP_006723077.1 | ||||
XM_011526452.2 | Intron | XP_011524754.1 | ||||
XM_011526453.2 | Intron | XP_011524755.1 |