Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGGTGGTATAGTGAAATAAAATT[A/T]AAAAGGAGTTGAAGACCTGTATCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608533 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXO38 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FBXO38 - F-box protein 38 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271723.1 | Intron | NP_001258652.1 | ||||
NM_030793.4 | Intron | NP_110420.3 | ||||
XM_005268513.1 | Intron | XP_005268570.1 | ||||
XM_006714797.1 | Intron | XP_006714860.1 | ||||
XM_011537684.2 | Intron | XP_011535986.1 | ||||
XM_017009899.1 | Intron | XP_016865388.1 | ||||
XM_017009900.1 | Intron | XP_016865389.1 | ||||
XM_017009901.1 | Intron | XP_016865390.1 | ||||
XM_017009902.1 | Intron | XP_016865391.1 |
LOC102546294 - uncharacterized LOC102546294 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |