Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATGCATGGAGTGGACCTGTAGGC[A/G]ACTTGCATCGTCTTCAACATGAAGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605010 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPINK5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SPINK5 - serine peptidase inhibitor, Kazal type 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127698.1 | 38 | UTR 5 | NP_001121170.1 | |||
NM_001127699.1 | 38 | UTR 5 | NP_001121171.1 | |||
NM_006846.3 | 38 | UTR 5 | NP_006837.2 | |||
XM_011537551.2 | 38 | UTR 5 | XP_011535853.1 |
Set Membership: |
HapMap |