Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTCCCCATAACCCTGGCCCCTGA[C/T]ACAAGGCCCGACCATCTAGACCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600327 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
SYT3 - synaptotagmin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160328.1 | Intron | NP_001153800.1 | ||||
NM_001160329.1 | Intron | NP_001153801.1 | ||||
NM_032298.2 | Intron | NP_115674.1 | ||||
XM_011527390.2 | Intron | XP_011525692.1 | ||||
XM_011527391.2 | Intron | XP_011525693.1 |
Set Membership: |
HapMap Validated |