Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606639 MIM: 605090 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GFM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GFM1 - G elongation factor mitochondrial 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RARRES1 - retinoic acid receptor responder 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002888.3 | Intron | NP_002879.2 | ||||
NM_206963.1 | Intron | NP_996846.1 | ||||
XM_005247686.4 | Intron | XP_005247743.3 |
Set Membership: |
HapMap |