Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 108729 MIM: 601720 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP5C1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ATP5C1 - ATP synthase, H+ transporting, mitochondrial F1 complex, gamma polypeptide 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001973.2 | Intron | NP_001001973.1 | ||||
NM_001320886.1 | Intron | NP_001307815.1 | ||||
NM_005174.3 | Intron | NP_005165.1 | ||||
XM_017016290.1 | Intron | XP_016871779.1 |
KIN - Kin17 DNA and RNA binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012311.3 | Intron | NP_036443.1 | ||||
XM_006717434.3 | Intron | XP_006717497.1 | ||||
XM_006717435.3 | Intron | XP_006717498.1 | ||||
XM_011519427.1 | Intron | XP_011517729.1 | ||||
XM_017015946.1 | Intron | XP_016871435.1 |
Set Membership: |
HapMap Validated |