Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGATGTTTGTGGTTGCTTACAATT[C/T]TCAATAGACACCACTTGCCACCCAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610606 MIM: 610637 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPEB3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CPEB3 - cytoplasmic polyadenylation element binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178137.1 | Intron | NP_001171608.1 | ||||
NM_014912.4 | Intron | NP_055727.3 |
MARCH5 - membrane associated ring-CH-type finger 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017824.4 | Intron | NP_060294.1 | ||||
XM_011539886.2 | Intron | XP_011538188.1 |
Set Membership: |
HapMap |