Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCACCTGAACTAGACCTTGAAAGA[C/T]GGGAACTCCAGGTGAAGAATGAGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
68 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610278 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PEAR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PEAR1 - platelet endothelial aggregation receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080471.1 | 1064 | Intron | NP_001073940.1 | |||
XM_005245141.3 | 1064 | Intron | XP_005245198.1 | |||
XM_011509510.2 | 1064 | Intron | XP_011507812.1 | |||
XM_011509511.2 | 1064 | Intron | XP_011507813.1 | |||
XM_011509512.2 | 1064 | Intron | XP_011507814.1 | |||
XM_017001234.1 | 1064 | UTR 5 | XP_016856723.1 | |||
XM_017001235.1 | 1064 | UTR 5 | XP_016856724.1 | |||
XM_017001236.1 | 1064 | Intron | XP_016856725.1 | |||
XM_017001237.1 | 1064 | Intron | XP_016856726.1 | |||
XM_017001238.1 | 1064 | Intron | XP_016856727.1 | |||
XM_017001239.1 | 1064 | Intron | XP_016856728.1 | |||
XM_017001240.1 | 1064 | Intron | XP_016856729.1 | |||
XM_017001241.1 | 1064 | Intron | XP_016856730.1 | |||
XM_017001242.1 | 1064 | Intron | XP_016856731.1 |
Set Membership: |
HapMap |