Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609104 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXO34 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBXO34 - F-box protein 34 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017943.3 | 36 | Intron | NP_060413.2 | |||
NM_152231.1 | 36 | UTR 5 | NP_689417.1 | |||
XM_005267791.4 | 36 | Intron | XP_005267848.1 | |||
XM_006720185.3 | 36 | Intron | XP_006720248.1 | |||
XM_017021391.1 | 36 | UTR 5 | XP_016876880.1 | |||
XM_017021392.1 | 36 | Intron | XP_016876881.1 | |||
XM_017021393.1 | 36 | Intron | XP_016876882.1 |