Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAAAAAAGAAAGATGGAATTCTA[C/T]GATATGTATTCTTCTGTGTCTGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TXNDC8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TXNDC8 - thioredoxin domain containing 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003936.3 | Intron | NP_001003936.1 | ||||
NM_001286946.1 | Intron | NP_001273875.1 | ||||
NM_001286947.1 | Intron | NP_001273876.1 | ||||
XM_011518463.2 | Intron | XP_011516765.1 | ||||
XM_017014579.1 | Intron | XP_016870068.1 | ||||
XM_017014580.1 | Intron | XP_016870069.1 | ||||
XM_017014581.1 | Intron | XP_016870070.1 | ||||
XM_017014582.1 | Intron | XP_016870071.1 | ||||
XM_017014583.1 | Intron | XP_016870072.1 | ||||
XM_017014584.1 | Intron | XP_016870073.1 |