Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTAATTATGCAAATTGATAATATT[A/T]GAGTTGCTTCCAAGAGCAACTAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613605 MIM: 602775 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BBIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BBIP1 - BBSome interacting protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPL13AP6 - ribosomal protein L13a pseudogene 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SHOC2 - SHOC2, leucine rich repeat scaffold protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001269039.2 | Intron | NP_001255968.1 | ||||
NM_001324336.1 | Intron | NP_001311265.1 | ||||
NM_001324337.1 | Intron | NP_001311266.1 | ||||
NM_007373.3 | Intron | NP_031399.2 | ||||
XM_017016702.1 | Intron | XP_016872191.1 | ||||
XM_017016703.1 | Intron | XP_016872192.1 | ||||
XM_017016704.1 | Intron | XP_016872193.1 |