Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTCAGCGCTGCTGGAGGATGAGG[C/T]CCTCATGTCTATGGCTCCCACCCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612944 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNASET2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RNASET2 - ribonuclease T2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003730.4 | Intron | NP_003721.2 | ||||
XM_017011397.1 | Intron | XP_016866886.1 | ||||
XM_017011398.1 | Intron | XP_016866887.1 | ||||
XM_017011399.1 | Intron | XP_016866888.1 |