Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGTAATTGTTAGAAAGTTTTGCA[A/G]TGGAGTGTATAGCTAACAAGCTCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
40 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602171 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DUSP27 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
DUSP27 - dual specificity phosphatase 27 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080426.1 | Intron | NP_001073895.1 | ||||
XM_005245603.3 | Intron | XP_005245660.1 | ||||
XM_011510146.2 | Intron | XP_011508448.1 |
GPA33 - glycoprotein A33 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105371598 - uncharacterized LOC105371598 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |