Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGCCAGTAGGGCCTCTGTGCAGC[C/T]TTGGCATGTTTCCTCTCCACAGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610223 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RIN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RIN3 - Ras and Rab interactor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319987.1 | Intron | NP_001306916.1 | ||||
NM_024832.4 | Intron | NP_079108.3 | ||||
XM_011537164.2 | Intron | XP_011535466.1 | ||||
XM_011537165.2 | Intron | XP_011535467.1 | ||||
XM_017021651.1 | Intron | XP_016877140.1 | ||||
XM_017021652.1 | Intron | XP_016877141.1 | ||||
XM_017021653.1 | Intron | XP_016877142.1 | ||||
XM_017021654.1 | Intron | XP_016877143.1 |