Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615077 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TBC1D30 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
TBC1D30 - TBC1 domain family member 30 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015279.1 | Intron | NP_056094.1 | ||||
XM_011538078.2 | Intron | XP_011536380.1 | ||||
XM_017019083.1 | Intron | XP_016874572.1 | ||||
XM_017019084.1 | Intron | XP_016874573.1 | ||||
XM_017019085.1 | Intron | XP_016874574.1 | ||||
XM_017019086.1 | Intron | XP_016874575.1 | ||||
XM_017019087.1 | Intron | XP_016874576.1 |
Set Membership: |
HapMap Validated |