Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCCCGGTGCACCCCCCTTTGCAC[A/G]CTGTCCGCTCCCTAAGGTCGCTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615258 MIM: 604723 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
METTL21B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
METTL21B - methyltransferase like 21B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015433.2 | Intron | NP_056248.2 | ||||
NM_206914.1 | Intron | NP_996797.1 |
TSFM - Ts translation elongation factor, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172695.1 | Intron | NP_001166166.1 | ||||
NM_001172696.1 | Intron | NP_001166167.1 | ||||
NM_001172697.1 | Intron | NP_001166168.1 | ||||
NM_005726.5 | Intron | NP_005717.3 |