Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGCCCAACTTTCTGCTGCCTCCT[C/G]GTGGCCACATTTCAACACTGCAACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605423 MIM: 610007 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DHH PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DHH - desert hedgehog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LMBR1L - limb development membrane protein 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300750.1 | Intron | NP_001287679.1 | ||||
NM_001300751.1 | Intron | NP_001287680.1 | ||||
NM_018113.3 | Intron | NP_060583.2 | ||||
XM_011538563.2 | Intron | XP_011536865.1 | ||||
XM_011538564.2 | Intron | XP_011536866.1 | ||||
XM_011538565.2 | Intron | XP_011536867.1 | ||||
XM_011538566.2 | Intron | XP_011536868.1 | ||||
XM_017019626.1 | Intron | XP_016875115.1 | ||||
XM_017019627.1 | Intron | XP_016875116.1 | ||||
XM_017019628.1 | Intron | XP_016875117.1 | ||||
XM_017019629.1 | Intron | XP_016875118.1 | ||||
XM_017019630.1 | Intron | XP_016875119.1 | ||||
XM_017019631.1 | Intron | XP_016875120.1 |
Set Membership: |
Validated |