Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRIQ1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LRRIQ1 - leucine rich repeats and IQ motif containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079910.1 | 302 | Missense Mutation | TAT,TGT | Y,C 83 | NP_001073379.1 | |
XM_011538817.2 | 302 | Missense Mutation | TAT,TGT | Y,C 83 | XP_011537119.1 | |
XM_011538818.2 | 302 | Missense Mutation | TAT,TGT | Y,C 83 | XP_011537120.1 | |
XM_011538819.2 | 302 | Missense Mutation | TAT,TGT | Y,C 83 | XP_011537121.1 | |
XM_011538823.2 | 302 | Missense Mutation | TAT,TGT | Y,C 83 | XP_011537125.1 | |
XM_011538824.2 | 302 | Missense Mutation | TAT,TGT | Y,C 83 | XP_011537126.1 | |
XM_011538825.2 | 302 | Missense Mutation | TAT,TGT | Y,C 83 | XP_011537127.1 | |
XM_017020019.1 | 302 | Missense Mutation | TAT,TGT | Y,C 83 | XP_016875508.1 |
TSPAN19 - tetraspanin 19 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |