Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGTGCTGCTGGTGGAGAGAACTGA[C/T]ATCTAACGCATGTGGCCCAGGAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NIM1K PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NIM1K - NIM1 serine/threonine protein kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153361.3 | Intron | NP_699192.1 | ||||
XM_006714450.1 | Intron | XP_006714513.1 | ||||
XM_006714451.1 | Intron | XP_006714514.1 | ||||
XM_017009164.1 | Intron | XP_016864653.1 | ||||
XM_017009165.1 | Intron | XP_016864654.1 |
Set Membership: |
HapMap |