Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCTCCTTCCCCACGCCAGCCCCT[C/T]GCTCCGGATTGGTCCTTCAGGAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
NIM1K PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NIM1K - NIM1 serine/threonine protein kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153361.3 | 82 | UTR 5 | NP_699192.1 | |||
XM_006714450.1 | 82 | Intron | XP_006714513.1 | |||
XM_006714451.1 | 82 | UTR 5 | XP_006714514.1 | |||
XM_017009164.1 | 82 | UTR 5 | XP_016864653.1 | |||
XM_017009165.1 | 82 | Intron | XP_016864654.1 |