Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACCGATGTATGCCAGGAATCCAG[A/G]CAGAGGGTGACATTAGCACTTCTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NIM1K PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
NIM1K - NIM1 serine/threonine protein kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153361.3 | Intron | NP_699192.1 | ||||
XM_006714450.1 | Intron | XP_006714513.1 | ||||
XM_006714451.1 | Intron | XP_006714514.1 | ||||
XM_017009164.1 | Intron | XP_016864653.1 | ||||
XM_017009165.1 | Intron | XP_016864654.1 |
Set Membership: |
HapMap Validated |