Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATACTAAATATATTTGGATTTTA[C/T]ACTTAAATAGGTCGGTTTTATGCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600067 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UGT2B4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
UGT2B4 - UDP glucuronosyltransferase family 2 member B4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297615.1 | Intron | NP_001284544.1 | ||||
NM_001297616.1 | Intron | NP_001284545.1 | ||||
NM_021139.2 | Intron | NP_066962.2 |