Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATACCCTCTATGATTTCCAAACAC[A/T]CACAGGAACATATACACCCAAAACA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602502 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GOLGA1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GOLGA1 - golgin A1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCAI - suppressor of cancer cell invasion | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144877.2 | 7031 | UTR 3 | NP_001138349.1 | |||
NM_173690.4 | 7031 | UTR 3 | NP_775961.2 |