Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATGAGAGATTTTCAGCCAATTGT[A/T]GGGTCAGAGCCCCAGAGTGAATGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603698 MIM: 602669 | ||||||||||||||||||||
Literature Links: |
GBF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GBF1 - golgi brefeldin A resistant guanine nucleotide exchange factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199378.1 | Intron | NP_001186307.1 | ||||
NM_001199379.1 | Intron | NP_001186308.1 | ||||
NM_004193.2 | Intron | NP_004184.1 | ||||
XM_005270261.1 | Intron | XP_005270318.1 | ||||
XM_006718047.1 | Intron | XP_006718110.1 | ||||
XM_006718048.1 | Intron | XP_006718111.1 | ||||
XM_006718049.1 | Intron | XP_006718112.1 | ||||
XM_006718050.1 | Intron | XP_006718113.1 | ||||
XM_011540312.1 | Intron | XP_011538614.1 | ||||
XM_011540313.1 | Intron | XP_011538615.1 | ||||
XM_011540314.2 | Intron | XP_011538616.1 | ||||
XM_017016861.1 | Intron | XP_016872350.1 | ||||
XM_017016862.1 | Intron | XP_016872351.1 | ||||
XM_017016863.1 | Intron | XP_016872352.1 | ||||
XM_017016864.1 | Intron | XP_016872353.1 |
PITX3 - paired like homeodomain 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |