Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611815 MIM: 603698 MIM: 602669 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELOVL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ELOVL3 - ELOVL fatty acid elongase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GBF1 - golgi brefeldin A resistant guanine nucleotide exchange factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PITX3 - paired like homeodomain 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005029.3 | Intron | NP_005020.1 |
Set Membership: |
HapMap |