Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608669 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BNC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
BNC2 - basonuclin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317939.1 | 15794 | UTR 3 | NP_001304868.1 | |||
NM_001317940.1 | 15794 | UTR 3 | NP_001304869.1 | |||
NM_017637.5 | 15794 | UTR 3 | NP_060107.3 | |||
XM_011517924.1 | 15794 | UTR 3 | XP_011516226.1 | |||
XM_011517934.2 | 15794 | Intron | XP_011516236.1 | |||
XM_017014816.1 | 15794 | UTR 3 | XP_016870305.1 | |||
XM_017014817.1 | 15794 | UTR 3 | XP_016870306.1 | |||
XM_017014818.1 | 15794 | UTR 3 | XP_016870307.1 | |||
XM_017014819.1 | 15794 | Intron | XP_016870308.1 | |||
XM_017014820.1 | 15794 | UTR 3 | XP_016870309.1 | |||
XM_017014821.1 | 15794 | Intron | XP_016870310.1 | |||
XM_017014822.1 | 15794 | UTR 3 | XP_016870311.1 | |||
XM_017014823.1 | 15794 | UTR 3 | XP_016870312.1 | |||
XM_017014824.1 | 15794 | UTR 3 | XP_016870313.1 | |||
XM_017014825.1 | 15794 | UTR 3 | XP_016870314.1 | |||
XM_017014826.1 | 15794 | Intron | XP_016870315.1 | |||
XM_017014827.1 | 15794 | Intron | XP_016870316.1 | |||
XM_017014828.1 | 15794 | Intron | XP_016870317.1 | |||
XM_017014829.1 | 15794 | UTR 3 | XP_016870318.1 |
Set Membership: |
HapMap Validated |