Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TUBGCP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TUBGCP3 - tubulin gamma complex associated protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286277.1 | Intron | NP_001273206.1 | ||||
NM_001286278.1 | Intron | NP_001273207.1 | ||||
NM_001286279.1 | Intron | NP_001273208.1 | ||||
NM_006322.5 | Intron | NP_006313.1 | ||||
XM_005268293.1 | Intron | XP_005268350.1 | ||||
XM_011537462.2 | Intron | XP_011535764.1 | ||||
XM_011537463.2 | Intron | XP_011535765.1 | ||||
XM_011537464.2 | Intron | XP_011535766.1 | ||||
XM_017020323.1 | Intron | XP_016875812.1 |
Set Membership: |
HapMap |