Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTCTTTCTGAAATTGCTCTGACA[C/T]ATCCAGTCCAAGCATAAGAGAGACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613109 MIM: 608332 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GM2A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GM2A - GM2 ganglioside activator | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC36A3 - solute carrier family 36 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145017.1 | Intron | NP_001138489.1 | ||||
NM_181774.3 | Intron | NP_861439.3 | ||||
XM_011537627.2 | Intron | XP_011535929.1 | ||||
XM_011537629.2 | Intron | XP_011535931.1 | ||||
XM_011537630.1 | Intron | XP_011535932.1 | ||||
XM_011537631.1 | Intron | XP_011535933.1 | ||||
XM_011537633.1 | Intron | XP_011535935.1 | ||||
XM_011537634.2 | Intron | XP_011535936.1 |
Set Membership: |
HapMap |