Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAAGGAAGAGTGGGGAGGCAGGA[C/T]GGGGCACAGGTGAGCTGGGACAAGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610097 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ODF4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ODF4 - outer dense fiber of sperm tails 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319953.1 | 282 | Missense Mutation | CGG,TGG | R,W 34 | NP_001306882.1 | |
NM_153007.4 | 282 | Missense Mutation | CGG,TGG | R,W 34 | NP_694552.2 | |
XM_005256476.3 | 282 | UTR 5 | XP_005256533.1 | |||
XM_011523688.1 | 282 | Intron | XP_011521990.1 |
Set Membership: |
HapMap |