Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608504 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGEF15 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARHGEF15 - Rho guanine nucleotide exchange factor 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025014.1 | 268 | Silent Mutation | ACC,ACT | T,T 59 | NP_079290.1 | |
NM_173728.3 | 268 | Silent Mutation | ACC,ACT | T,T 59 | NP_776089.2 | |
XM_011523734.1 | 268 | Silent Mutation | ACC,ACT | T,T 59 | XP_011522036.1 | |
XM_011523735.1 | 268 | Silent Mutation | ACC,ACT | T,T 59 | XP_011522037.1 | |
XM_011523736.1 | 268 | Silent Mutation | ACC,ACT | T,T 59 | XP_011522038.1 | |
XM_011523737.1 | 268 | Intron | XP_011522039.1 |
Set Membership: |
HapMap |