Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
93 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OR6N1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
OR6N1 - olfactory receptor family 6 subfamily N member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005185.1 | 813 | Missense Mutation | ACA,ATA | T,I 194 | NP_001005185.1 | |
XM_017000325.1 | 813 | Missense Mutation | ACA,ATA | T,I 194 | XP_016855814.1 | |
XM_017000326.1 | 813 | Missense Mutation | ACA,ATA | T,I 194 | XP_016855815.1 | |
XM_017000327.1 | 813 | Missense Mutation | ACA,ATA | T,I 194 | XP_016855816.1 |
Set Membership: |
HapMap Validated |