Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGATCTCAGCTTTTAGTTGGCCAA[G/T]AATATATTTAAGAATTGGGCCACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607030 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GCA PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
GCA - grancalcin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005246446.2 | Intron | XP_005246503.1 | ||||
XM_006712398.3 | Intron | XP_006712461.1 | ||||
XM_006712400.3 | Intron | XP_006712463.1 | ||||
XM_006712401.3 | Intron | XP_006712464.1 | ||||
XM_011510926.2 | Intron | XP_011509228.1 | ||||
XM_011510927.2 | Intron | XP_011509229.1 | ||||
XM_011510928.2 | Intron | XP_011509230.1 | ||||
XM_017003764.1 | Intron | XP_016859253.1 | ||||
XM_017003765.1 | Intron | XP_016859254.1 | ||||
XM_017003766.1 | Intron | XP_016859255.1 | ||||
XM_017003767.1 | Intron | XP_016859256.1 | ||||
XM_017003768.1 | Intron | XP_016859257.1 | ||||
XM_017003769.1 | Intron | XP_016859258.1 |