Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACATATATATAATTTCTCAAGAAG[C/T]TTTCATTGAGTCCAAATTGCTTAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607030 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GCA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GCA - grancalcin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005246446.2 | Intron | XP_005246503.1 | ||||
XM_006712398.3 | Intron | XP_006712461.1 | ||||
XM_006712400.3 | Intron | XP_006712463.1 | ||||
XM_006712401.3 | Intron | XP_006712464.1 | ||||
XM_011510926.2 | Intron | XP_011509228.1 | ||||
XM_011510927.2 | Intron | XP_011509229.1 | ||||
XM_011510928.2 | Intron | XP_011509230.1 | ||||
XM_017003764.1 | Intron | XP_016859253.1 | ||||
XM_017003765.1 | Intron | XP_016859254.1 | ||||
XM_017003766.1 | Intron | XP_016859255.1 | ||||
XM_017003767.1 | Intron | XP_016859256.1 | ||||
XM_017003768.1 | Intron | XP_016859257.1 | ||||
XM_017003769.1 | Intron | XP_016859258.1 |