Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGGGATCAGCAGCTGGGGCTCCT[G/C]CTTCAAACCAGGGCCCCCATGCTCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602166 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP3B2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AP3B2 - adaptor related protein complex 3 beta 2 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278511.1 | Intron | NP_001265440.1 | ||||
NM_001278512.1 | Intron | NP_001265441.1 | ||||
NM_004644.4 | Intron | NP_004635.2 | ||||
XM_017022640.1 | Intron | XP_016878129.1 | ||||
XM_017022641.1 | Intron | XP_016878130.1 |
CPEB1-AS1 - CPEB1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |