Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608025 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NBAS PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NBAS - neuroblastoma amplified sequence | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015909.3 | 7058 | Intron | NP_056993.2 | |||
XM_011510357.2 | 7058 | Intron | XP_011508659.1 | |||
XM_011510358.2 | 7058 | Intron | XP_011508660.1 | |||
XM_011510359.2 | 7058 | Intron | XP_011508661.1 | |||
XM_011510360.2 | 7058 | Intron | XP_011508662.1 | |||
XM_011510361.2 | 7058 | Intron | XP_011508663.1 | |||
XM_017004317.1 | 7058 | UTR 3 | XP_016859806.1 |