Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTCTTTCTGAAGGATAGGGACAA[C/T]ATTTTACTCATCTTAGTATCCTTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MCMDC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MCMDC2 - minichromosome maintenance domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136160.1 | Intron | NP_001129632.1 | ||||
NM_001136161.1 | Intron | NP_001129633.1 | ||||
NM_173518.4 | Intron | NP_775789.3 | ||||
XM_005251174.2 | Intron | XP_005251231.1 | ||||
XM_006716427.3 | Intron | XP_006716490.1 | ||||
XM_006716429.2 | Intron | XP_006716492.1 | ||||
XM_006716433.3 | Intron | XP_006716496.1 | ||||
XM_011517467.2 | Intron | XP_011515769.1 | ||||
XM_011517468.2 | Intron | XP_011515770.1 | ||||
XM_011517469.2 | Intron | XP_011515771.1 | ||||
XM_017013140.1 | Intron | XP_016868629.1 |
Set Membership: |
HapMap |