Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605702 MIM: 604673 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LYVE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LYVE1 - lymphatic vessel endothelial hyaluronan receptor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRVI1 - murine retrovirus integration site 1 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098579.2 | 3870 | UTR 3 | NP_001092049.2 | |||
NM_001100163.2 | 3870 | UTR 3 | NP_001093633.1 | |||
NM_001100167.2 | 3870 | UTR 3 | NP_001093637.1 | |||
NM_001206880.1 | 3870 | UTR 3 | NP_001193809.1 | |||
NM_001206881.1 | 3870 | UTR 3 | NP_001193810.1 | |||
NM_130385.3 | 3870 | UTR 3 | NP_569056.4 |
MRVI1-AS1 - MRVI1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |