Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGTGTATTTACACCTAGGCTATA[C/T]GGTGTAACCTACTGCTCCTAGGCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600788 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TCEB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
TCEB1 - transcription elongation factor B subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204857.1 | Intron | NP_001191786.1 | ||||
NM_001204858.1 | Intron | NP_001191787.1 | ||||
NM_001204859.1 | Intron | NP_001191788.1 | ||||
NM_001204860.1 | Intron | NP_001191789.1 | ||||
NM_001204861.1 | Intron | NP_001191790.1 | ||||
NM_001204862.1 | Intron | NP_001191791.1 | ||||
NM_001204863.1 | Intron | NP_001191792.1 | ||||
NM_001204864.1 | Intron | NP_001191793.1 | ||||
NM_005648.3 | Intron | NP_005639.1 | ||||
XM_011517580.2 | Intron | XP_011515882.1 | ||||
XM_011517581.2 | Intron | XP_011515883.1 |
Set Membership: |
HapMap |