Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616642 MIM: 601301 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C6orf89 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C6orf89 - chromosome 6 open reading frame 89 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286635.1 | Intron | NP_001273564.1 | ||||
NM_001286636.1 | Intron | NP_001273565.1 | ||||
NM_001286637.1 | Intron | NP_001273566.1 | ||||
NM_152734.3 | Intron | NP_689947.2 | ||||
XM_011514377.2 | Intron | XP_011512679.1 | ||||
XM_017010432.1 | Intron | XP_016865921.1 | ||||
XM_017010433.1 | Intron | XP_016865922.1 | ||||
XM_017010434.1 | Intron | XP_016865923.1 |
PPIL1 - peptidylprolyl isomerase like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |