Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAACCTCAGAGCCTCCCCTGTGC[A/G]AGGCTGAGTTCATCACAAGGATTCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604177 MIM: 194526 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6850 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
MIR6850 - microRNA 6850 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPL8 - ribosomal protein L8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000973.4 | Intron | NP_000964.1 | ||||
NM_001317771.1 | Intron | NP_001304700.1 | ||||
NM_001317782.1 | Intron | NP_001304711.1 | ||||
NM_033301.2 | Intron | NP_150644.1 | ||||
XM_017013723.1 | Intron | XP_016869212.1 |
ZNF34 - zinc finger protein 34 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF517 - zinc finger protein 517 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |