Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKFN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKFN1 - ankyrin repeat and fibronectin type III domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153228.2 | Intron | NP_694960.2 | ||||
XM_006721728.3 | Intron | XP_006721791.1 | ||||
XM_011524428.2 | Intron | XP_011522730.1 | ||||
XM_011524429.2 | Intron | XP_011522731.1 | ||||
XM_011524430.2 | Intron | XP_011522732.1 | ||||
XM_011524431.2 | Intron | XP_011522733.1 | ||||
XM_011524433.2 | Intron | XP_011522735.1 | ||||
XM_011524434.2 | Intron | XP_011522736.1 | ||||
XM_017024263.1 | Intron | XP_016879752.1 | ||||
XM_017024264.1 | Intron | XP_016879753.1 | ||||
XM_017024265.1 | Intron | XP_016879754.1 | ||||
XM_017024266.1 | Intron | XP_016879755.1 | ||||
XM_017024267.1 | Intron | XP_016879756.1 | ||||
XM_017024268.1 | Intron | XP_016879757.1 | ||||
XM_017024269.1 | Intron | XP_016879758.1 | ||||
XM_017024270.1 | Intron | XP_016879759.1 |