Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGCAGCAGCAGCAACAGCAACTG[A/C]GAAACGTAAGTGAGAACTAAGTCGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601638 MIM: 607388 MIM: 605493 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARFIP2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARFIP2 - ADP ribosylation factor interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242854.1 | 104 | Intron | NP_001229783.1 | |||
NM_001242855.1 | 104 | Intron | NP_001229784.1 | |||
NM_001242856.1 | 104 | Intron | NP_001229785.1 | |||
NM_012402.3 | 104 | Intron | NP_036534.1 | |||
XM_005252840.4 | 104 | Intron | XP_005252897.1 |
TIMM10B - translocase of inner mitochondrial membrane 10 homolog B (yeast) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012192.3 | 104 | Silent Mutation | AGA,CGA | R,R 12 | NP_036324.1 |
TRIM3 - tripartite motif containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |