Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTAGGGGGGATTATAAAACCTAAC[A/C]GAGGATAATGAAGGCCTCCTTACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYNPO2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SYNPO2 - synaptopodin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128933.2 | Intron | NP_001122405.1 | ||||
NM_001128934.2 | Intron | NP_001122406.1 | ||||
NM_001286754.1 | Intron | NP_001273683.1 | ||||
NM_001286755.1 | Intron | NP_001273684.1 | ||||
NM_133477.2 | Intron | NP_597734.2 |
Set Membership: |
HapMap |