Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAGGGAGCCTGATGAGGGATGAA[C/T]AGCAATGCCCTGAATTCCAAATGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602192 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAM10 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ADAM10 - ADAM metallopeptidase domain 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001110.3 | Intron | NP_001101.1 | ||||
NM_001320570.1 | Intron | NP_001307499.1 | ||||
XM_017021861.1 | Intron | XP_016877350.1 |
HSP90AB4P - heat shock protein 90 alpha family class B member 4, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
Validated |